Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichlorobenzothiazole 95%

Supplier: AMBEED, INC
Warning

A128062-1G A128062-250MG A128062-1G
103801-060EA 23 USD InStock
103801-060 76971-944 77313-348
2,4-Dichlorobenzothiazole 95%
2,4-Dichlorobenzothiazole
Precaución: for Research Use Only


Formula: C₇H₃Cl₂NS
Boiling Pt: 286.8 °C (760 mm Hg)
Melting Pt: 97 °C
Flash Pt: 127.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00127716
CAS Number: 3622-30-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty