Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloropyrimidine-5-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A172958-1G A172958-1G A172958-5G A172958-10G A172958-250MG A172958-25G
103806-772EA 12 USD InStock
103806-772 103845-064 77082-288 77082-290 77248-084 77248-086 77301-236
2,4-Dichloropyrimidine-5-carbonitrile 95%
2,4-Dichloropyrimidine-5-carbonitrile
Precaución: for Research Use Only


Formula: C₅HCl₂N₃
Boiling Pt: 323.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09746248
CAS Number: 3177-24-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty