Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Diaminobenzonitrile 97%

Supplier: AMBEED, INC
Warning

A306987-1G A306987-5G A306987-10G A306987-250MG A306987-1G A306987-5G
103814-492EA 29 USD InStock
103814-492 103835-666 77141-630 77141-632 77305-946 77305-948
2,5-Diaminobenzonitrile 97%
2,5-Diaminobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₇N₃
Boiling Pt: 371 °C (760 mm Hg)
Melting Pt: 83…87 °C
Flash Pt: 178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077966
CAS Number: 14346-13-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty