Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromobenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A281038-1G A281038-5G A281038-100G A281038-10G A281038-250MG A281038-1G A281038-25G A281038-5G
103813-532EA 15 USD InStock
103813-532 103853-484 76647-940 76647-942 76647-944 76837-176 76837-178 76837-180
2,5-Dibromobenzaldehyde 98%
2,5-Dibromobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₄Br₂O
Melting Pt: 88…92 °C
Flash Pt: 111.244 °C
Storage Temperature: Ambient
MDL Number: MFCD00462864
CAS Number: 74553-29-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty