Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromophenol 98%

Supplier: AMBEED, INC
Warning

A124634-1G A124634-10G A124634-250MG A124634-25G A124634-1G A124634-5G
103800-562EA 8 USD InStock
103800-562 77246-260 77246-262 77246-264 77312-698 77312-700
2,5-Dibromophenol 98%
2,5-Dibromophenol
Precaución: for Research Use Only


Formula: C₆H₄Br₂O
Flash Pt: 101.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01851373
CAS Number: 28165-52-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty