Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-2,5-dibromopyridine 98%

Supplier: AMBEED, INC
Warning

A178478-1G A178478-1G A178478-5G A178478-10G A178478-250MG
103807-342EA 6 USD InStock
103807-342 103845-678 77083-112 77083-114 77248-196 77248-198
3-Amino-2,5-dibromopyridine 98%
3-Amino-2,5-dibromopyridine
Precaución: for Research Use Only


Formula: C₅H₄Br₂N₂
Boiling Pt: 325 °C (760 mm Hg)
Flash Pt: 150.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09266224
CAS Number: 90902-84-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty