Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichlorobenzoxazole 98%

Supplier: AMBEED, INC
Warning

A139721-1G A139721-10G A139721-1G A139721-5G A139721-100G
103802-764EA 8 USD InStock
103802-764 76972-532 77299-086 77315-860 77670-996
2,5-Dichlorobenzoxazole 98%
2,5-Dichlorobenzoxazole
Precaución: for Research Use Only


Formula: C₇H₃Cl₂NO
Melting Pt: 43…47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09264312
CAS Number: 3621-81-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty