Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloroquinoline 95%

Supplier: AMBEED, INC
Warning

A158751-1G A158751-5G A158751-100MG A158751-250MG A158751-1G
103805-166EA 45 USD InStock
103805-166 103843-380 76984-962 76984-964 77319-234
2,5-Dichloroquinoline 95%
2,5-Dichloroquinoline
Precaución: for Research Use Only


Formula: C₉H₅Cl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD00832074
CAS Number: 59412-12-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty