Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dihydroxyterephthalaldehyde 95%

Supplier: AMBEED, INC
Warning

A856742-100MG A856742-250MG A856742-5G
103841-252EA 211 USD InStock
103841-252 76820-772 76820-774 76845-510
2,5-Dihydroxyterephthalaldehyde 95%
2,5-Dihydroxyterephthalaldehyde
Precaución: for Research Use Only


Formula: C₈H₆O₄
Melting Pt: 262 °C at N/A mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD22055335
CAS Number: 1951-36-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty