Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethylbenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A354797-5G A354797-1G A354797-10G A354797-25G A354797-5G
103826-130EA 8 USD InStock
103826-130 103837-134 76840-262 77251-592 77306-668 77306-670
2,5-Dimethylbenzaldehyde 95%
2,5-Dimethylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₀O
Boiling Pt: 217.1 °C (760 mm Hg)
Flash Pt: 87.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003342
CAS Number: 5779-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty