Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethylpyridin-4-amine 98%

Supplier: AMBEED, INC
Warning

A124163-1G A124163-100MG A124163-250MG
103800-492EA 74 USD InStock
103800-492 76983-376 76983-378
2,5-Dimethylpyridin-4-amine 98%
2,5-Dimethylpyridin-4-amine
Precaución: for Research Use Only


Formula: C₇H₁₀N₂
MW: 122.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235163
CAS Number: 22279-89-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty