Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethylterephthalonitrile 97%

Supplier: AMBEED, INC
Warning

A206551-1G A206551-5G A206551-250MG A206551-25G A206551-1G A206551-5G
103817-712EA 40 USD InStock
103817-712 103848-388 77248-972 77248-974 77302-990 77302-992
2,5-Dimethylterephthalonitrile 97%
2,5-Dimethylterephthalonitrile
Precaución: for Research Use Only


Formula: C₁₀H₈N₂
Storage Temperature: Ambient
MDL Number: MFCD00191411
CAS Number: 39095-25-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty