Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Succinimidyl 6-Biotinamidohexanoate 98%

Supplier: AMBEED, INC
Warning

A140101-1G A140101-100MG A140101-5G
103802-832EA 268 USD InStock
103802-832 77624-168 77876-971
N-Succinimidyl 6-Biotinamidohexanoate 98%
N-Succinimidyl 6-Biotinamidohexanoate
Precaución: Inert atmosphere, store in freezer, under –20 °C.


Formula: C₂₀H₃₀N₄O₆S
Melting Pt: 169…171 °C
Storage Temperature: Freezer
MDL Number: MFCD00065502
CAS Number: 72040-63-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty