Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromo-p-cresol 98%

Supplier: AMBEED, INC
Warning

A248977-5G A248977-25G A248977-100G A248977-10G A248977-5G
103851-684EA 14 USD InStock
103851-684 77091-028 77249-940 77249-942 77304-468
2,6-Dibromo-p-cresol 98%
2,6-Dibromo-p-cresol
Precaución: for Research Use Only


Formula: C₇H₆Br₂O
MW: 265.93
Boiling Pt: 235.8 °C (760 mm Hg)
Melting Pt: 2
Density: 2.22
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002154
CAS Number: 2432-14-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty