Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromoisonicotinonitrile 95%

Supplier: AMBEED, INC
Warning

A184028-1G A184028-5G A184028-100MG A184028-250MG
103815-702EA 51 USD InStock
103815-702 103846-262 77163-960 77163-962
2,6-Dibromoisonicotinonitrile 95%
2,6-Dibromoisonicotinonitrile
Precaución: Inert atmosphere, 2…8 °C.


Formula: C₆H₂Br₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD03086034
CAS Number: 408352-58-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty