Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3-hydroxyisonicotinic acid 98%

Supplier: AMBEED, INC
Warning

A135269-1G A135269-250MG A135269-1G
103802-124EA 344 USD InStock
103802-124 77246-700 77298-766
2,6-Dichloro-3-hydroxyisonicotinic acid 98%
2,6-Dichloro-3-hydroxyisonicotinic acid
Precaución: for Research Use Only


Formula: C₆H₃Cl₂NO₃
MW: 207.9989 g/mol
Boiling Pt: 548.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03411718
CAS Number: 185422-96-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty