Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3-nitropyridin-4-amine 97%

Supplier: AMBEED, INC
Warning

A183707-1G A183707-5G A183707-1G A183707-25G A183707-5G A183707-100G A183707-10G A183707-250MG
103815-662EA 11 USD InStock
103815-662 103846-224 77083-828 77083-830 77083-832 77248-352 77248-354 77248-356
2,6-Dichloro-3-nitropyridin-4-amine 97%
2,6-Dichloro-3-nitropyridin-4-amine
Precaución: for Research Use Only


Formula: C₅H₃Cl₂N₃O₂
MW: 208.0022 g/mol
Melting Pt: 148…150 °C
Flash Pt: 211.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03265754
CAS Number: 2897-43-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty