Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-4-fluorobenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A130632-1G A130632-10G A130632-250MG A130632-1G A130632-5G
103801-444EA 19 USD InStock
103801-444 76870-704 76870-706 77313-894 77313-896
2,6-Dichloro-4-fluorobenzaldehyde 95%
2,6-Dichloro-4-fluorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₃Cl₂FO
MW: 193.003 g/mol
Boiling Pt: 235.5±35.0 °C (760 mm Hg)
Flash Pt: 96.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20526366
CAS Number: 1182709-86-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty