Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-4-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A221677-1G A221677-5G A221677-1G A221677-10G A221677-500G A221677-25G
103818-942EA 5 USD InStock
103818-942 103849-820 77088-418 77249-402 77249-404 77303-610
2,6-Dichloro-4-methylpyridine 98%
2,6-Dichloro-4-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 241.3 °C (760 mm Hg)
Flash Pt: 123 °C
Storage Temperature: Freezer
MDL Number: MFCD09264302
CAS Number: 39621-00-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty