Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-7-methylpurine 97%

Supplier: AMBEED, INC
Warning

A127856-1G A127856-250MG A127856-1G A127856-25G
103801-026EA 26 USD InStock
103801-026 77094-850 77298-238 77313-302
2,6-Dichloro-7-methylpurine 97%
2,6-Dichloro-7-methylpurine
Precaución: for Research Use Only


Formula: C₆H₄Cl₂N₄
Boiling Pt: 307.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00039690
CAS Number: 2273-93-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty