Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3-pyridineboronic acid 97%

Supplier: AMBEED, INC
Warning

A144174-1G A144174-5G A144174-100G A144174-10G A144174-250MG A144174-1G A144174-25G A144174-5G
103803-370EA 19 USD InStock
103803-370 103841-448 76984-360 76984-362 76984-364 77316-684 77316-686 77316-688
2,6-Dichloro-3-pyridineboronic acid 97%
2,6-Dichloro-3-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₅H₄BCl₂NO₂
MW: 191.81
Boiling Pt: 373.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03094989
CAS Number: 148493-34-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty