Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloronicotinonitrile 97%

Supplier: AMBEED, INC
Danger

A143247-1G A143247-10G A143247-250MG A143247-1G A143247-25G A143247-5G
103803-256EA 9 USD InStock
103803-256 77095-504 77095-506 77299-332 77316-506 77316-508
2,6-Dichloronicotinonitrile 97%
2,6-Dichloronicotinonitrile
Precaución: for Research Use Only


Formula: C₆H₂Cl₂N₂
MW: 173.00 g/mol
Boiling Pt: 286 °C (760 mm Hg)
Melting Pt: 121…124 °C
Storage Temperature: Ambient
MDL Number: MFCD03411341
CAS Number: 40381-90-6
UN: 3439
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty