Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-3-nitropyridine 97%

Supplier: AMBEED, INC
Warning

A408543-5G A408543-1G A408543-100G A408543-10G A408543-250MG A408543-25G A408543-5G
103827-188EA 11 USD InStock
103827-188 103838-234 76841-748 76876-950 76876-952 76876-954 76876-956 77307-130
2,6-Difluoro-3-nitropyridine 97%
2,6-Difluoro-3-nitropyridine
Precaución: for Research Use Only


Formula: C₅H₂F₂N₂O₂
MW: 160.078 g/mol
Melting Pt: 6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11044323
CAS Number: 58602-02-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty