Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-4-methoxybenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A278876-1G A278876-5G A278876-25G A278876-5G A278876-100G A278876-10G A278876-1G
103813-434EA 9 USD InStock
103813-434 103853-378 77093-304 77093-306 77238-842 77238-844 77305-308
2,6-Difluoro-4-methoxybenzaldehyde 98%
2,6-Difluoro-4-methoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₆F₂O₂
Boiling Pt: 207.1 °C (760 mm Hg)
Melting Pt: 73…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114368
CAS Number: 256417-10-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty