Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoroaniline 98%

Supplier: AMBEED, INC
Danger

A101896-25G A101896-5G
103807-824EA 237 USD InStock
103807-824 77282-858 77282-860
2,6-Difluoroaniline 98%
2,6-Difluoroaniline
Precaución: for Research Use Only


Formula: C₆H₅F₂N
Boiling Pt: 154 °C (760 mm Hg)
Flash Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007655
CAS Number: 5509-65-9
UN: 1992
ADR: 3,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty