Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diiodopyrazine 97%

Supplier: AMBEED, INC
Warning

A113178-1G A113178-250MG A113178-1G
103798-178EA 37 USD InStock
103798-178 103824-020 76781-228 77310-846
2,6-Diiodopyrazine 97%
2,6-Diiodopyrazine
Precaución: for Research Use Only


Formula: C₄H₂I₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD09265492
CAS Number: 58138-79-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty