Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Xylidine hydrochloride 96%

Supplier: AMBEED, INC
Warning

A246020-1G A246020-5G A246020-1G A246020-25G A246020-5G A246020-100G
103811-682EA 6 USD InStock
103811-682 103851-512 77090-788 77090-790 77090-792 77249-880
2,6-Xylidine hydrochloride 96%
2,6-Xylidine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₂ClN
Storage Temperature: Ambient
MDL Number: MFCD00060213
CAS Number: 21436-98-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty