Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dimethylbenzonitrile 98%

Supplier: AMBEED, INC
Warning

A162557-1G A162557-5G A162557-10G A162557-1G
103805-626EA 5 USD InStock
103805-626 103843-876 77273-386 77319-924
2,6-Dimethylbenzonitrile 98%
2,6-Dimethylbenzonitrile
Precaución: for Research Use Only


Formula: C₉H₉N
MW: 131.18
Boiling Pt: 228.7 °C (760 mm Hg)
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00037166
CAS Number: 6575-13-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty