Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dimethylhydroquinone 97%

Supplier: AMBEED, INC
Danger

A105654-1G A105654-5G A105654-100G A105654-250MG A105654-25G A105654-1G A105654-5G
103796-920EA 9 USD InStock
103796-920 103822-658 77245-406 77245-408 77245-410 77283-372 77283-374
2,6-Dimethylhydroquinone 97%
2,6-Dimethylhydroquinone
Precaución: for Research Use Only


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 284.6 °C (760 mm Hg)
Melting Pt: 154 °C
Flash Pt: 141.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00016466
CAS Number: 654-42-2
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty