Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Di-tert-butyl-4-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A105364-1G A105364-5G A105364-100G A105364-10G A105364-250MG A105364-25G A105364-500G A105364-1G A105364-5G
103796-878EA 10 USD InStock
103796-878 103822-606 77093-728 77093-730 77093-732 77093-734 77093-736 77309-476 77309-478
2,6-Di-tert-butyl-4-methylpyridine 98%
2,6-Di-tert-butyl-4-methylpyridine
Precaución: for Research Use Only


Formula: C₁₄H₂₃N
MW: 205.34 g/mol
Boiling Pt: 233 °C (760 mm Hg)
Melting Pt: 33…36 °C
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006305
CAS Number: 38222-83-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty