Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Pyridinedimethanol 98%

Supplier: AMBEED, INC
Warning

A146017-1G A146017-5G A146017-100G A146017-10G A146017-500G A146017-1G A146017-25G A146017-5G
103803-618EA 5 USD InStock
103803-618 103841-704 77190-606 77190-608 77190-610 77299-492 77299-494 77299-496
2,6-Pyridinedimethanol 98%
2,6-Pyridinedimethanol
Precaución: for Research Use Only


Formula: C₇H₉NO₂
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00006351
CAS Number: 1195-59-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty