Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,8-Diazaspiro[4.5]decan-3-one 95%

Supplier: AMBEED, INC
Warning

A148160-1G A148160-5G A148160-100MG A148160-250MG
103803-880EA 89 USD InStock
103803-880 103841-994 76972-936 76972-938
2,8-Diazaspiro[4.5]decan-3-one 95%
2,8-Diazaspiro[4.5]decan-3-one
Precaución: for Research Use Only


Formula: C₈H₁₄N₂O
Boiling Pt: 360.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11044206
CAS Number: 561314-57-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty