Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,9-Dichloro-1,10-phenanthroline 95%

Supplier: AMBEED, INC
Warning

A184088-5G A184088-1G A184088-5G A184088-100MG A184088-10G A184088-250MG
103815-710EA 28 USD InStock
103815-710 103846-270 77083-882 77083-884 77248-368 77248-370 77248-372
2,9-Dichloro-1,10-phenanthroline 95%
2,9-Dichloro-1,10-phenanthroline
Precaución: for Research Use Only


Formula: C₁₂H₆Cl₂N₂
Melting Pt: 249…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02084186
CAS Number: 29176-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty