Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allylboronic acid pinacol ester 97% stabilised

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with phenothiazine

A115496-1G A115496-10G A115496-25G A115496-1G A115496-5G
103798-576EA 9 USD InStock
103798-576 103824-432 77257-926 77257-928 77297-374 77311-250
Allylboronic acid pinacol ester 97% stabilised
Allylboronic acid pinacol ester


Formula: C₉H₁₇BO₂
Boiling Pt: 131.6 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00013347
CAS Number: 72824-04-5
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty