Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-α-(4-Hydroxyphenyl)glycine 97%

Supplier: AMBEED, INC
Warning

A111647-5G A111647-100G A111647-10G A111647-25G
103823-696EA 6 USD InStock
103823-696 76869-760 76869-762 76869-764
DL-α-(4-Hydroxyphenyl)glycine 97%
DL-α-(4-Hydroxyphenyl)glycine
Precaución: for Research Use Only


Formula: C₈H₉NO₃
MW: 167.1600 g/mol
Boiling Pt: 446.3 °C (760 mm Hg)
Melting Pt: 240 °C
Flash Pt: 223.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00065931
CAS Number: 938-97-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty