Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-DL-tyrosine 97%

Supplier: AMBEED, INC
Warning

A151995-1G A151995-100MG A151995-1G A151995-5G
103804-334EA 173 USD InStock
103804-334 76689-390 77318-082 77625-186
3-Fluoro-DL-tyrosine 97%
3-Fluoro-DL-tyrosine
Precaución: for Research Use Only


Formula: C₉H₁₀FNO₃
MW: 199.18 g/mol
Melting Pt: 280 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063075
CAS Number: 403-90-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty