Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromoanthranilic acid 97%

Supplier: AMBEED, INC
Warning

A102883-1G A102883-5G A102883-10G A102883-25G A102883-500G A102883-1G A102883-5G
103796-438EA 9 USD InStock
103796-438 103822-140 77135-568 77135-570 77135-572 77283-010 77309-012
3-Bromoanthranilic acid 97%
3-Bromoanthranilic acid
Precaución: for Research Use Only


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 170…172 °C
Flash Pt: 161.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03618453
CAS Number: 20776-51-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty