Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-bromobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A597346-5G A597346-1G A597346-100G A597346-10G A597346-250MG A597346-25G A597346-5G
103828-960EA 8 USD InStock
103828-960 103840-098 76844-248 77005-364 77005-366 77005-368 77307-914 77307-916
2-Amino-3-bromobenzonitrile 98%
2-Amino-3-bromobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₅BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD05670539
CAS Number: 114344-60-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty