Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-chloropyridine 98%

Supplier: AMBEED, INC
Warning

A430401-5G A430401-25G A430401-100G A430401-10G A430401-1G A430401-500G A430401-5G
103838-570EA 9 USD InStock
103838-570 76842-256 77016-570 77016-572 77016-574 77016-576 77307-246
2-Amino-3-chloropyridine 98%
2-Amino-3-chloropyridine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂
Boiling Pt: 311.5 °C (760 mm Hg)
Melting Pt: 177…178 °C
Storage Temperature: Ambient
MDL Number: MFCD03541052
CAS Number: 39620-04-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty