Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitroanthranilic acid 98%

Supplier: AMBEED, INC
Warning

A132299-100G A132299-10G A132299-500G A132299-5G A132299-25G
77136-936EA 86.52 USD InStock
77136-936 77136-938 77136-940 77298-552 77314-280
3-Nitroanthranilic acid 98%
3-Nitroanthranilic acid
Precaución: for Research Use Only


Formula: C₇H₆N₂O₄
MW: 182.14
Boiling Pt: 390.4 °C (760 mm Hg)
Melting Pt: 207…211 °C
Flash Pt: 189.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00024261
CAS Number: 606-18-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty