Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A228058-1G A228058-5G A228058-100G A228058-10G A228058-250MG A228058-25G A228058-1G
103819-356EA 11 USD InStock
103819-356 103850-264 77000-116 77000-118 77000-120 77089-054 77303-814
2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile 95%
2-Amino-4,5-dimethyl-1H-pyrrole-3-carbonitrile
Precaución: for Research Use Only


Formula: C₇H₉N₃
Boiling Pt: 366.2 °C (760 mm Hg)
Melting Pt: 178…181 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00849263
CAS Number: 21392-51-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty