Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,6-dimethoxybenzamide 97%

Supplier: AMBEED, INC
Warning

A436294-1G A436294-5G A436294-10G A436294-250MG
103827-610EA 110 USD InStock
103827-610 103838-674 76842-406 76842-408 76877-322 76877-324
2-Amino-4,6-dimethoxybenzamide 97%
2-Amino-4,6-dimethoxybenzamide
Precaución: for Research Use Only


Formula: C₉H₁₂N₂O₃
MW: 196.2 g/mol
Boiling Pt: 331.6±42.0 °C (760 mm Hg)
Flash Pt: 165.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20040374
CAS Number: 63920-73-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty