Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,6-dimethoxypyrimidine 97%

Supplier: AMBEED, INC
Warning

A489915-100G A489915-10G A489915-500G A489915-25G
103810-384EA 73 USD InStock
103810-384 77279-592 77279-594 77279-596 77307-582
2-Amino-4,6-dimethoxypyrimidine 97%
2-Amino-4,6-dimethoxypyrimidine
Precaución: for Research Use Only


Formula: C₆H₉N₃O₂
Boiling Pt: 356.4 °C (760 mm Hg)
Melting Pt: 94…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038832
CAS Number: 36315-01-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty