Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,6-dimethylpyridine 97%

Supplier: AMBEED, INC
Warning

A258499-1G A258499-5G A258499-1G A258499-25G A258499-5G A258499-100G A258499-10G
103812-376EA 6 USD InStock
103812-376 103852-254 77091-780 77091-782 77091-784 77113-624 77113-626
2-Amino-4,6-dimethylpyridine 97%
2-Amino-4,6-dimethylpyridine
Precaución: for Research Use Only


Formula: C₇H₁₀N₂
MW: 122.1700 g/mol
Boiling Pt: 235 °C (760 mm Hg)
Melting Pt: 63…64 °C
Flash Pt: 115.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00006322
CAS Number: 5407-87-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty