Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromoanthranilic acid 97%

Supplier: AMBEED, INC
Warning

A156798-1G A156798-5G A156798-100G A156798-10G A156798-500G A156798-1G A156798-5G A156798-25G
103804-922EA 6 USD InStock
103804-922 103843-124 77137-986 77137-988 77137-990 77300-214 77300-216 77318-924
4-Bromoanthranilic acid 97%
4-Bromoanthranilic acid
Precaución: for Research Use Only


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 352.4 °C (760 mm Hg)
Melting Pt: 230…234 °C
Flash Pt: 166.9 °C
Storage Temperature: Ambient
MDL Number: MFCD03618454
CAS Number: 20776-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty