Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-bromopyridine 98%

Supplier: AMBEED, INC
Warning

A267871-1G A267871-5G A267871-100G A267871-10G A267871-500G A267871-25G A267871-5G A267871-1G
103812-884EA 6 USD InStock
103812-884 103852-784 76874-982 76874-984 76874-986 77092-534 77092-536 77304-992
2-Amino-4-bromopyridine 98%
2-Amino-4-bromopyridine
Precaución: for Research Use Only


Formula: C₅H₅BrN₂
Melting Pt: 142…146 °C
Flash Pt: 159.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01646115
CAS Number: 84249-14-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty