Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloroanthranilic acid 95%

Supplier: AMBEED, INC
Warning

A161735-1000G A161735-100G A161735-10G A161735-500G A161735-25G
103808-898EA 137 USD InStock
103808-898 77138-182 77138-184 77138-186 77319-774
4-Chloroanthranilic acid 95%
4-Chloroanthranilic acid


Formula: C₇H₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00007778
CAS Number: 89-77-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty