Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxyanthranilic acid 98%

Supplier: AMBEED, INC
Warning

A141753-1G A141753-100G A141753-10G A141753-500G A141753-1G A141753-25G A141753-5G
103803-054EA 9 USD InStock
103803-054 77162-344 77162-346 77162-348 77316-230 77316-232 77316-234
4-Methoxyanthranilic acid 98%
4-Methoxyanthranilic acid
Precaución: for Research Use Only


Formula: C₈H₉NO₃
MW: 167.16
Boiling Pt: 339.8 °C (760 mm Hg)
Melting Pt: 174…177 °C
Flash Pt: 159.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00667729
CAS Number: 4294-95-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty