Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-methylpyrimidine-5-carbonitrile 98%

Supplier: AMBEED, INC
Warning

A281773-1G A281773-5G A281773-1G A281773-100MG
103813-572EA 290 USD InStock
103813-572 103853-524 76837-224 77114-090
2-Amino-4-methylpyrimidine-5-carbonitrile 98%
2-Amino-4-methylpyrimidine-5-carbonitrile
Precaución: for Research Use Only


Formula: C₆H₆N₄
Boiling Pt: 386.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09833141
CAS Number: 17321-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty