Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-nitrobenzamide 97%

Supplier: AMBEED, INC
Warning

A165668-1G A165668-5G A165668-10G A165668-1G A165668-5G
103805-984EA 29 USD InStock
103805-984 103844-240 77247-800 77320-460 77320-462
2-Amino-4-nitrobenzamide 97%
2-Amino-4-nitrobenzamide
Precaución: for Research Use Only


Formula: C₇H₇N₃O₃
MW: 181.1500 g/mol
Boiling Pt: 432.2 °C (760 mm Hg)
Flash Pt: 189.388 °C
Storage Temperature: Ambient
MDL Number: MFCD07368613
CAS Number: 31930-18-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty